Assay Details
| Gene Symbol: MAP2K4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022418585.1 | XP_022274293.1 | 4 - 5 | 570 | 103 | |
| XM_022418586.1 | XP_022274294.1 | 4 - 5 | 588 | 103 | ||
More Information
Set Membership: |
|

MAP2K4
Genomic Map