Assay Details
| Gene Symbol: TEAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410963.1 | XP_022266671.1 | 8 - 9 | 1333 | 182 | |
| XM_022410964.1 | XP_022266672.1 | 7 - 8 | 1199 | 182 | ||
| XM_022410965.1 | XP_022266673.1 | 8 - 9 | 1335 | 182 | ||
More Information
Set Membership: |
|

TEAD4
Genomic Map