Assay Details
| Gene Symbol: VBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005642049.3 | XP_005642106.2 | 4 - 5 | 315 | 103 | |
| XM_538210.6 | XP_538210.1 | 4 - 5 | 450 | 103 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

VBP1
Genomic Map