Assay Details
| Gene Symbol: VIPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022403936.1 | XP_022259644.1 | 6 - 7 | 692 | 95 | |
| XM_022403937.1 | XP_022259645.1 | 5 - 6 | 608 | 95 | ||
| XM_849361.4 | XP_854454.2 | 6 - 7 | 715 | 95 | ||
More Information
Set Membership: |
|

VIPR2
Genomic Map