Assay Details
| Gene Symbol: WIPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640331.3 | XP_005640388.1 | 7 - 8 | 1709 | 142 | |
| XM_005640332.3 | XP_005640389.1 | 7 - 8 | 1590 | 142 | ||
| XM_545531.6 | XP_545531.3 | 7 - 8 | 1641 | 142 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

WIPF1
Genomic Map