Assay Details
| Gene Symbol: ZNF76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014118385.2 | XP_013973860.1 | 6 - 7 | 730 | 96 | |
| XM_014118386.2 | XP_013973861.1 | 6 - 7 | 836 | 96 | ||
| XM_014118387.2 | XP_013973862.1 | 6 - 7 | 730 | 96 | ||
| XM_538875.6 | XP_538875.2 | 6 - 7 | 949 | 96 | ||
More Information
Set Membership: |
|

ZNF76
Genomic Map