Assay Details
| Gene Symbol: AIF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625228.3 | XP_005625285.1 | 7 | 506 | 74 | |
| XM_005625231.3 | XP_005625288.1 | 5 | 443 | 74 | ||
| XM_014116996.2 | XP_013972471.1 | 5 | 437 | 74 | ||
| XM_014116997.2 | XP_013972472.1 | 4 | 1814 | 74 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AIF1L
Genomic Map