Assay Details
| Gene Symbol: SERHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625795.3 | XP_005625852.1 | 2 | 142 | 88 | |
| XM_014117303.2 | XP_013972778.1 | 2 | 142 | 88 | ||
| XM_022424345.1 | XP_022280053.1 | 2 | 142 | 88 | ||
| XM_022424346.1 | XP_022280054.1 | 2 | 264 | 88 | ||
| XM_022424347.1 | XP_022280055.1 | 2 | 194 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022424346.1, XM_022424347.1 transcripts.
Set Membership: |
|

SERHL2
Genomic Map