Assay Details
| Gene Symbol: KCTD15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022426203.1 | XP_022281911.1 | 3 - 4 | 207 | 130 | |
| XM_022426214.1 | XP_022281922.1 | 3 - 4 | 292 | 130 | ||
| XM_541709.5 | XP_541709.2 | 2 - 3 | 264 | 130 | ||
More Information
Set Membership: |
|

KCTD15
Genomic Map