Assay Details
| Gene Symbol: ZMAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640141.3 | XP_003640189.1 | 2 - 3 | 491 | 108 | |
| XM_022414143.1 | XP_022269851.1 | 2 - 3 | 491 | 108 | ||
| XM_022414144.1 | XP_022269852.1 | 3 - 4 | 685 | 108 | ||
| XM_022414145.1 | XP_022269853.1 | 4 - 5 | 797 | 108 | ||
| XM_022414146.1 | XP_022269854.1 | 2 - 3 | 427 | 108 | ||
More Information
Set Membership: |
|

ZMAT3
Genomic Map