Assay Details
| Gene Symbol: SLC16A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637517.2 | XP_005637574.1 | 6 | 1200 | 119 | |
| XM_005637518.2 | XP_005637575.1 | 7 | 1484 | 119 | ||
| XM_005637520.2 | XP_005637577.1 | 6 | 1484 | 119 | ||
| XM_005637521.2 | XP_005637578.1 | 6 | 1452 | 119 | ||
| XM_005637522.3 | XP_005637579.1 | 6 | 1200 | 119 | ||
| XM_005637523.3 | XP_005637580.1 | 6 | 1401 | 119 | ||
| XM_005637524.3 | XP_005637581.1 | 6 | 1270 | 119 | ||
| XM_005637525.2 | XP_005637582.1 | 5 | 1056 | 119 | ||
| XM_005637526.2 | XP_005637583.1 | 6 | 1200 | 119 | ||
More Information
Set Membership: |
|

SLC16A12
Genomic Map