Assay Details
| Gene Symbol: LIPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623126.3 | XP_005623183.1 | 3 - 4 | 489 | 135 | |
| XM_022421668.1 | XP_022277376.1 | 3 - 4 | 2031 | 135 | ||
More Information
Set Membership: |
|

LIPG
Genomic Map