Assay Details
| Gene Symbol: LHFPL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627264.3 | XP_005627321.1 | 1 | 456 | 102 | |
| XM_022425971.1 | XP_022281679.1 | 1 | 462 | 102 | ||
More Information
Set Membership: |
|

LHFPL5
Genomic Map