Assay Details
| Gene Symbol: VAPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635226.3 | XP_005635283.1 | 4 - 5 | 486 | 126 | |
| XM_022408827.1 | XP_022264535.1 | 4 - 5 | 432 | 126 | ||
| XM_534466.6 | XP_534466.2 | 4 - 5 | 600 | 126 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

VAPB
Genomic Map