Assay Details
| Gene Symbol: YOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622310.3 | XP_005622367.1 | 482 | 135 | ||
| XM_547387.5 | XP_547387.1 | 1215 | 135 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: YOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622310.3 | XP_005622367.1 | 482 | 135 | ||
| XM_547387.5 | XP_547387.1 | 1215 | 135 | |||
Set Membership: |
|