Assay Details
| Gene Symbol: LRCH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014114211.2 | XP_013969686.1 | 1964 | 113 | ||
| XM_014114212.2 | XP_013969687.1 | 1778 | 113 | |||
| XM_022419638.1 | XP_022275346.1 | 1715 | 113 | |||
| XM_022419639.1 | XP_022275347.1 | 1841 | 113 | |||
| XM_022419640.1 | XP_022275348.1 | 1818 | 113 | |||
| XM_022419641.1 | XP_022275349.1 | 1800 | 113 | |||
| XM_546954.7 | XP_546954.5 | 1964 | 113 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LRCH4
Genomic Map