Assay Details
| Gene Symbol: SP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022411232.1 | XP_022266940.1 | 2 | 663 | 150 | |
| XM_844688.5 | XP_849781.2 | 3 | 2043 | 150 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022411232.1 | XP_022266940.1 | 2 | 663 | 150 | |
| XM_844688.5 | XP_849781.2 | 3 | 2043 | 150 | ||
Set Membership: |
|