Assay Details
| Gene Symbol: ZNF184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110503.2 | XP_013965978.1 | 6 | 2330 | 110 | |
| XM_014110504.2 | XP_013965979.1 | 6 | 2197 | 110 | ||
| XM_014110505.2 | XP_013965980.1 | 6 | 2378 | 110 | ||
More Information
Set Membership: |
|

ZNF184
Genomic Map