Assay Details
| Gene Symbol: TM6SF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618397.3 | XP_005618454.1 | 3 - 4 | 356 | 95 | |
| XM_022416983.1 | XP_022272691.1 | 3 - 4 | 366 | 95 | ||
| XM_022416984.1 | XP_022272692.1 | 3 - 4 | 366 | 95 | ||
| XM_022416985.1 | XP_022272693.1 | 3 - 4 | 366 | 95 | ||
| XM_022416986.1 | XP_022272694.1 | 3 - 4 | 214 | 95 | ||
| XM_022416987.1 | XP_022272695.1 | 2 - 3 | 221 | 95 | ||
| XM_022416990.1 | XP_022272698.1 | 3 - 4 | 368 | 95 | ||
| XM_022416991.1 | XP_022272699.1 | 3 - 4 | 387 | 95 | ||
| XM_536207.7 | XP_536207.4 | 3 - 4 | 353 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022416986.1, XM_022416987.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TM6SF1
Genomic Map