Assay Details
| Gene Symbol: CKMT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413031.1 | XP_022268739.1 | 7 - 8 | 1153 | 154 | |
| XM_535451.6 | XP_535451.1 | 6 - 7 | 1102 | 154 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CKMT1A
Genomic Map