Assay Details
| Gene Symbol: NGDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115771.2 | XP_013971246.1 | 6 - 7 | 624 | 174 | |
| XM_022421816.1 | XP_022277524.1 | 6 - 7 | 924 | 174 | ||
| XM_547734.6 | XP_547734.2 | 6 - 7 | 552 | 174 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NGDN
Genomic Map