Assay Details
| Gene Symbol: DISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111486.2 | XP_013966961.1 | 19 | 3697 | 103 | |
| XM_022414600.1 | XP_022270308.1 | 19 | 3727 | 103 | ||
| XM_845529.5 | XP_850622.2 | 19 | 3727 | 103 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DISP3
Genomic Map