Assay Details
| Gene Symbol: MARCHF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022403562.1 | XP_022259270.1 | 6 - 7 | 1231 | 153 | |
| XM_022403564.1 | XP_022259272.1 | 5 - 6 | 540 | 153 | ||
| XM_022403565.1 | XP_022259273.1 | 3 - 4 | 719 | 153 | ||
| XM_022403566.1 | XP_022259274.1 | 2 - 3 | 404 | 153 | ||
More Information
Set Membership: |
|

MARCHF1
Genomic Map