Assay Details
| Gene Symbol: BCL2L13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637386.3 | XP_005637443.1 | 3 | 420 | 169 | |
| XM_005637387.3 | XP_005637444.1 | 3 | 383 | 169 | ||
| XM_005637393.3 | XP_005637450.1 | 3 | 420 | 169 | ||
| XM_014108379.2 | XP_013963854.1 | 3 | 420 | 169 | ||
| XM_014108380.2 | XP_013963855.1 | 3 | 420 | 169 | ||
| XM_534939.6 | XP_534939.3 | 3 | 419 | 169 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

BCL2L13
Genomic Map