Assay Details
| Gene Symbol: LRRC39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621872.3 | XP_005621929.1 | 5 - 6 | 736 | 135 | |
| XM_022420432.1 | XP_022276140.1 | 5 - 6 | 743 | 135 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LRRC39
Genomic Map