Assay Details
Gene Symbol: PCDH20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005633953.3 | XP_005634010.1 | 2 | 2067 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
