Assay Details
| Gene Symbol: SEPTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625820.3 | XP_005625877.1 | 10 - 11 | 1150 | 106 | |
| XM_014117312.2 | XP_013972787.1 | 11 - 12 | 2553 | 106 | ||
| XM_014117313.2 | XP_013972788.1 | 11 - 12 | 2555 | 106 | ||
| XM_022424352.1 | XP_022280060.1 | 12 - 13 | 2829 | 106 | ||
| XM_843889.5 | XP_848982.2 | 10 - 11 | 1164 | 106 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SEPTIN3
Genomic Map