Assay Details
| Gene Symbol: TLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116391.1 | XP_013971866.1 | 1984 | 72 | ||
| XM_014116392.2 | XP_013971867.1 | 2228 | 72 | |||
| XM_014116393.2 | XP_013971868.1 | 2219 | 72 | |||
More Information
Set Membership: |
|

TLK2
Genomic Map