Assay Details
| Gene Symbol: SLC25A37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410097.1 | XP_022265805.1 | 6 | 757 | 187 | |
| XM_022410098.1 | XP_022265806.1 | 6 | 754 | 187 | ||
| XM_022410099.1 | XP_022265807.1 | 5 | 660 | 187 | ||
| XM_022410100.1 | XP_022265808.1 | 3 | 606 | 187 | ||
| XM_844354.5 | XP_849447.1 | 4 | 804 | 187 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC25A37
Genomic Map