Assay Details
| Gene Symbol: CTCFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014106851.2 | XP_013962326.1 | 8 - 9 | 1617 | 165 | |
| XM_022409197.1 | XP_022264905.1 | 8 - 9 | 1617 | 165 | ||
| XM_022409198.1 | XP_022264906.1 | 8 - 9 | 1618 | 165 | ||
| XM_022409199.1 | XP_022264907.1 | 8 - 9 | 1617 | 165 | ||
More Information
Set Membership: |
|

CTCFL
Genomic Map