Assay Details
| Gene Symbol: PTGS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003354.1 | NP_001003354.1 | 5 - 6 | 663 | 154 | |
| XM_005622227.3 | XP_005622284.1 | 5 - 6 | 690 | 154 | ||
| GenBank mRNA | AY044905.1 | - | 5 - 6 | 663 | 154 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PTGS2
Genomic Map