Assay Details
| Gene Symbol: DTX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620987.3 | XP_005621044.1 | 2374 | 157 | ||
| XM_005620988.3 | XP_005621045.1 | 2008 | 157 | |||
| XM_005620989.3 | XP_005621046.1 | 2425 | 157 | |||
| XM_014114179.2 | XP_013969654.1 | 2232 | 157 | |||
| XM_022419589.1 | XP_022275297.1 | 1800 | 157 | |||
| XM_022419590.1 | XP_022275298.1 | 1751 | 157 | |||
| XM_536849.5 | XP_536849.2 | 2209 | 157 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DTX2
Genomic Map