Assay Details
Gene Symbol: CCDC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636147.2 | XP_005636204.1 | 1083 | 92 | ||
XM_005636148.2 | XP_005636205.1 | 1080 | 92 | |||
XM_005636150.2 | XP_005636207.1 | 1086 | 92 | |||
XM_005636151.2 | XP_005636208.1 | 1014 | 92 | |||
XM_005636152.3 | XP_005636209.1 | 976 | 92 | |||
XM_014108087.2 | XP_013963562.1 | 1082 | 92 | |||
XM_014108088.2 | XP_013963563.1 | 981 | 92 | |||
XM_014108089.2 | XP_013963564.1 | 821 | 92 | |||
XM_022410379.1 | XP_022266087.1 | 1080 | 92 | |||
XM_022410380.1 | XP_022266088.1 | 1082 | 92 | |||
XM_022410382.1 | XP_022266090.1 | 981 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
