Assay Details
Gene Symbol: NECAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419117.1 | XP_022274825.1 | 8 - 9 | 2184 | 129 | |
XM_022419118.1 | XP_022274826.1 | 7 - 8 | 2169 | 129 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
