Assay Details
| Gene Symbol: TMEM54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617617.3 | XP_005617674.1 | 4 - 5 | 899 | 101 | |
| XM_022411823.1 | XP_022267531.1 | 4 - 5 | 769 | 101 | ||
| XM_849496.4 | XP_854589.2 | 4 - 5 | 589 | 101 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM54
Genomic Map