Assay Details
| Gene Symbol: EPRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640825.3 | XP_005640882.1 | 25 - 26 | 3582 | 84 | |
| XM_844375.5 | XP_849468.1 | 24 - 25 | 3560 | 84 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EPRS1
Genomic Map