Assay Details
| Gene Symbol: ZNF367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638869.4 | XP_003638917.1 | 1 - 2 | 539 | 136 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZNF367
Genomic Map