Assay Details
| Gene Symbol: MVB12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625352.2 | XP_005625409.1 | 3 - 4 | 359 | 130 | |
| XM_022423811.1 | XP_022279519.1 | 4 - 5 | 806 | 130 | ||
| XM_022423813.1 | XP_022279521.1 | 3 - 4 | 656 | 130 | ||
| XM_022423814.1 | XP_022279522.1 | 3 - 4 | 333 | 130 | ||
More Information
Set Membership: |
|

MVB12B
Genomic Map