Assay Details
| Gene Symbol: C7H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421373.1 | XP_022277081.1 | 1 - 2 | 410 | 195 | |
| XM_022421376.1 | XP_022277084.1 | 1 - 2 | 391 | 195 | ||
More Information
Set Membership: |
|

C7H1orf43
Genomic Map