Assay Details
| Gene Symbol: ANAPC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623994.3 | XP_005624051.1 | 630 | 172 | ||
| XM_022422623.1 | XP_022278331.1 | 747 | 289 | |||
| XM_022422624.1 | XP_022278332.1 | 420 | 289 | |||
| XM_540489.7 | XP_540489.2 | 297 | 172 | |||
More Information
Set Membership: |
|

ANAPC11
Genomic Map