Assay Details
| Gene Symbol: HNRNPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617817.3 | XP_005617874.1 | 6 - 7 | 840 | 69 | |
| XM_005617818.3 | XP_005617875.1 | 6 - 7 | 841 | 69 | ||
| XM_005617819.3 | XP_005617876.1 | 5 - 6 | 675 | 69 | ||
| XM_014110666.2 | XP_013966141.1 | 6 - 7 | 841 | 69 | ||
| XM_535365.6 | XP_535365.2 | 5 - 6 | 613 | 69 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HNRNPR
Genomic Map