Assay Details
| Gene Symbol: PHKG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621272.3 | XP_005621329.1 | 963 | 138 | ||
| XM_843877.5 | XP_848970.1 | 1079 | 138 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PHKG2
Genomic Map