Assay Details
| Gene Symbol: CDIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621600.3 | XP_005621657.1 | 7 | 883 | 172 | |
| XM_005621602.3 | XP_005621659.1 | 6 | 767 | 172 | ||
| XM_005621604.3 | XP_005621661.1 | 6 | 816 | 172 | ||
| XM_014114656.2 | XP_013970131.1 | 6 | 816 | 172 | ||
| XM_022420124.1 | XP_022275832.1 | 6 | 841 | 172 | ||
| XM_022420125.1 | XP_022275833.1 | 7 | 938 | 172 | ||
| XM_547145.6 | XP_547145.2 | 6 | 816 | 172 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDIP1
Genomic Map