Assay Details
| Gene Symbol: GMPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115787.2 | XP_013971262.1 | 2 - 3 | 301 | 130 | |
| XM_845034.4 | XP_850127.1 | 2 - 3 | 435 | 130 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GMPR2
Genomic Map