Assay Details
| Gene Symbol: JKAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623456.3 | XP_005623513.1 | 6 - 7 | 814 | 97 | |
| XM_022422058.1 | XP_022277766.1 | 8 - 9 | 1163 | 97 | ||
| XM_022422059.1 | XP_022277767.1 | 6 - 7 | 1082 | 97 | ||
| XM_547838.6 | XP_547838.4 | 6 - 7 | 842 | 97 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

JKAMP
Genomic Map