Assay Details
| Gene Symbol: TMEM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_536671.6 | XP_536671.2 | 2 | 746 | 112 | |
| XM_859742.5 | XP_864835.1 | 3 | 768 | 112 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM11
Genomic Map