Assay Details
| Gene Symbol: NMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624373.3 | XP_005624430.1 | 1621 | 65 | ||
| XM_022422885.1 | XP_022278593.1 | 1656 | 65 | |||
| XM_537613.5 | XP_537613.2 | 1517 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NMT1
Genomic Map