Assay Details
| Gene Symbol: PPARG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024632.2 | NP_001019803.1 | 895 | 127 | ||
| XM_005632011.3 | XP_005632068.1 | 931 | 127 | |||
| XM_005632012.3 | XP_005632069.1 | 940 | 127 | |||
| XM_005632014.3 | XP_005632071.1 | 806 | 127 | |||
| GenBank mRNA | DQ016141.3 | - | 895 | 127 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPARG
Genomic Map