Assay Details
| Gene Symbol: AKT1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616283.3 | XP_005616340.1 | 4 - 5 | 1124 | 132 | |
| XM_005616284.3 | XP_005616341.1 | 5 - 6 | 960 | 132 | ||
| XM_005616285.3 | XP_005616342.1 | 5 - 6 | 882 | 132 | ||
| XM_005616287.3 | XP_005616344.1 | 4 - 5 | 765 | 132 | ||
| XM_022423643.1 | XP_022279351.1 | 4 - 5 | 3022 | 132 | ||
| XM_022423645.1 | XP_022279353.1 | 5 - 6 | 2300 | 132 | ||
| XM_022423647.1 | XP_022279355.1 | 6 - 7 | 1264 | 132 | ||
| XM_022423650.1 | XP_022279358.1 | 4 - 5 | 908 | 132 | ||
More Information
Set Membership: |
|

AKT1S1
Genomic Map