Assay Details
| Gene Symbol: RAB25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313865.1 | NP_001300794.1 | 2 | 463 | 122 | |
| XM_014114978.2 | XP_013970453.1 | 2 | 506 | 122 | ||
| XM_022420762.1 | XP_022276470.1 | 2 | 511 | 122 | ||
More Information
Set Membership: |
|

RAB25
Genomic Map